Water purification system, arium® advance EDI

Leverancier: Sartorius
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H2O-EDI-2-T H2O-EDI-2-B H2O-EDI-1-T H2O-EDI-1-B
171-1296EA 8610 EUR
171-1296 VIVAH2O-EDI-2-B 171-1297 171-1298
Water purification system, arium® advance EDI
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
The arium® advance EDI provides Type 2 water in consistently high quality. The innovative iJust makes the best use of environmental resources by automatically optimizing water consumption. Unlike conventional water purification systems, the unique display with touch functions in combination with the intuitive menu navigation ensures easy operation.

  • Applied for microbiological media and reagents
  • Solutions for chemical analysis and synthesis, histology, ELISA, RIA and buffer solutions
  • Feed water for laboratory devices like autoclaves and dishwashers
  • Premium
More Product Information
Order Now

Specificaties

Model arium® advance EDI
Environmentally Preferable
Debiet =3 l/min
Weerstand bij +25 °C 5 - 15 MΩ.cm
Deeltje <1/ml (@ 0,2 μm)
Micro-organismen <1 CFU/1000 ml
b×d×h 350×451×501 mm
Gewicht in werking met volle tank 26 kg
Gewicht 19 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR