Waterdemineralisatiepatronen, Aquadem® SDF
Leverancier: ENVIROFALK PHARMAWATERSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1943-005 1943-015 1943-025
172-4151EA 1180 EUR
172-4151 172-4152 172-4153
Waterdemineralisatiepatronen, Aquadem® SDF
Waterzuiveringssystemen Deïonisatiesystemen
Pressure-resistant water deionisation cartridges made from stainless steel, filled with regenerable mixed-bed resins.
  • Economical water deionisation for all requirements
  • Achieved water quality covers the majority of common laboratory applications
  • Simple handling with quick connection system
  • Suitable for benchtop set up in laboratories
  • Long life time, transportable
Aquadem® SDF 20: Standard equipment for laboratory glass washers.
Bestelinformatie: De aansluitset dient afzonderlijk te worden besteld.