Reverse osmosis water purification system, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CDRC00201
172-5087EA 851 EUR
172-5087
Reverse osmosis water purification system, Millipore®
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
This Reverse Osmosis membrane removes 95 to 99% of contaminants including ions, particulates, bacteria, and organic molecules (MW >200kDa)

Height: 41 cm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR