Copper selenide ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

17901.06 17901.14
17901.06EA 70.1 EUR
17901.06 17901.14
Copper selenide ≥99.5% (metals basis)
Copper selenide

Purity: 99.5% CAS No.: 1317-41-5 Molecular Formula: CuSe Formula Weight: 142.50 Physical Form: 6mm & down pieces Density: 6.0-6.6 Melting Point: 550° dec. Solubility: Soluble in HCl with H2Se evolution, in H2SO4 with SO2 evolution. Oxidized to CuSeO3 by HNO3 Merck Reference: 14,2651 UN No.: 3077 MDL No.: MFCD00016058


In semiconductors, as catalyst in Kjeldahl digestion

Formule: CuSe
Dichtheid: 6,0…6.6
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016058
CAS nummer: 1317-41-5
EINECS: 215-272-8
UN: 3077
ADR: 9,III
Merck Index: 14,2651

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR