Inlet traps for rotary vane pumps
Leverancier: EDWARDS HIGH VACUUM METTLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A44410000 A44510000 A13305000
181-0096EA 0 EUR
181-0096 181-0097 181-0070
Inlet traps for rotary vane pumps
Vallen voor vacuümpompen
Inlet Desiccant Trap IDT20K: Use when pumping limited quantities of water vapour at high pumpings speeds to a low vapour pressure.
  • Inlet Chemical Trap ITC20K: Protects the pump against chemically active gases
  • Inlet Dust Filter ITF20K: Protects the pump against abrasive dust
Inlet Oil Vapour (foreline) Trap FL20K: Use on a clean pumping system to prevent backmigration of pump-oil vapour into the vacuum system.