Sulfamidezuur, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid

1828.0250EA 121.45 EUR
1828.0250 1828.1000
Sulfamidezuur, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sulfamidezuur
Formule: H₃NO₃S
MW: 97.09 g/mol
Kookpunt: 247
Smeltpunt: 205
Dichtheid: 2.15 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011603
CAS nummer: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8 III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min. 99.0 %
Chloride (Cl) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.005 %
Residue after Ignition max. 0.01 %
Sulfate (SO4) max. 0.02 %
Trace Impurities (in ppm) Iron (Fe) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR