Diaphragm valves, VM series
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20664010 20664011
183-1151EA 416 EUR
183-1151 183-1152
Diaphragm valves, VM series
Ventielen Vacuümventielen
VM series valves are hand operated diaphragm valves. They are used as isolation valves, air-admittance valves or manual-control valves.
  • Materials with high chemical resistance
  • High conductance, low leakage rate
  • Flow control possible
  • Gas-tight against atmosphere in both directions