Copper(I) bromide dimethyl sulphide complex

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

19368.04 19368.09 19368.18
19368.04EA 32.8 EUR
19368.04 19368.09 19368.18
Copper(I) bromide dimethyl sulphide complex
Copper(I) bromide dimethyl sulphide complex

CAS No.: 54678-23-8 Molecular Formula: CuBr.S(CH3)2 Formula Weight: 205.58 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: Pale brown or pale green Melting Point: 124-129° dec. UN No.: 3335 MDL No.: MFCD00043295

Formule: C₂H₆BrCuS
MW: 205.58 g/mol
Smeltpunt: 124…129 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043295
CAS nummer: 54678-23-8
EINECS: 259-292-5
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR