Butylbenzene, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: n-Butylbenzene

MQ100

19600-10ML
19600-10MLEA 213 EUR
19600-10ML
Butylbenzene, Supelco®
Butylbenzene
Formule: C₁₀H₁₄
MW: 134.22 g/mol
Kookpunt: 183
Smeltpunt: –88 °C
Dichtheid: 0.86 g/cm³ (20 °C)
Vlampunt: 61
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009463
CAS nummer: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3 III
Merck Index: 13,01548

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR