Benzamidine hydrochloride, Calbiochem®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

199001-25 199001-5
199001-25EA 130 EUR
199001-25 199001-5
Benzamidine hydrochloride, Calbiochem®
Benzamidine hydrochloride
Formule: C₇H₉ClN₂
Smeltpunt: 174…179 °C
MDL Nummer: MFCD00066285
CAS nummer: 1670-14-0
EINECS: 216-795-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR