Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20083 20083-1
20083.EA 359 EUR
20083. 20083-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)
Antilichamen
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR