Verbindingsstukken met conische slijping
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01501419 01501424 01501429 01501914 01501924 01501929 01501934 01502414 01502419 01502429 01502434 01502445 01502914 01502919 01502924 01502934 01502945 01502960 01503419 01503424 01503429 01504529 01501214 01501412 01502971 01504560 01507145 01504571 01506029 01507129 01504534 01506045
201-2115EA 14.2 EUR
201-2115 201-0094 201-2108 201-1100 201-2114 201-2105 201-2149 201-2103 201-2102 201-2110 201-2150 201-2151 201-2104 201-1104 271-2107 201-2113 201-2112 201-0629 201-0095 201-0096 201-2118 201-1105 201-2117 201-2148 201-2152 201-2153 201-2154 201-0631 201-0632 201-0634 201-0630 201-0633
Verbindingsstukken met conische slijping
Adapters Adapters met NS-verbindingen
DURAN®-borosilicaatglas.
  • Outstanding leak tightness
  • Minimal requirement for joint lubrication
  • Joints cannot be misaligned
Reduction adapter from large cone to small socket or expansion adapter from small cone to large socket. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12257
Certificaten: DIN 12257