Jood flessen met kraag en jood stop
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03026249
201-1162EA 73.4 EUR
201-1162
Jood flessen met kraag en jood stop
Kolven Jodium kolven
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.