Flasks, round bottom, narrow or wide neck, with rim
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632411203250_U 632411205966_U 632411203050 632411203100
201-1583EA 22.7 EUR
201-1583 201-1599 KAVA632411203050 KAVA632411203100
Flasks, round bottom, narrow or wide neck, with rim
Kolven Rondbodemkolven
Borosilicate glass 3,3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.
Order Now
Specificaties
Environmentally Preferable