Gasinlaatbuisjes met ventielafsluitkraan
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05086001
201-2612EA 89.1 EUR
201-2612
Gasinlaatbuisjes met ventielafsluitkraan
Buisjes Gasinlaatbuizen
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With valve stopcock for dosing, with glass olive (angle: 90°), with cone NS14/23, tube length from top of the joint: approx. 150 mm.