Koeler, dubbele oppervlakte met NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C5/12/SC C5/13/SC C5/22/SC C5/23/SC C5/24/SC C5/11/SC
201-8287EA 185 EUR
201-8287 201-8288 201-8289 201-8290 201-8291 201-8286
Koeler, dubbele oppervlakte met NS hals, Quickfit®
Condensors
PYREX®, borosilicaatglas.
  • Detachable plastic screw thread connector
  • High chemical and temperature resistance
Double surface, highly efficient condensers with a large cooling surface, inner cooling surface provides “baffle” effect whilst outer prevents “creep” with low boiling point liquids.