Chromotropic acid disodium salt dihydrate ≥98.0% ACS, Reag. Ph. Eur. analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

20261.132EA 113 EUR
20261.132
Chromotropic acid disodium salt dihydrate ≥98.0% ACS, Reag. Ph. Eur. analytical reagent
Chromotropic acid disodium salt dihydrate
Formule: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Smeltpunt: > 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150612
CAS nummer: 5808-22-0
EINECS: 204-972-9
Merck Index: 13,02261

Order Now

Specification Test Results

Assay (dried substance) Min. 98.0 %
Suited for formaldehyde reagent Passes test
Suited for titanium reagent Passes test
Water Max. 10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR