Tetranatriumethyleendiaminetetraacetaat ≥83%, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

20299.291EA 130 EUR
20299.291
Tetranatriumethyleendiaminetetraacetaat ≥83%, TECHNICAL
Tetranatriumethyleendiaminetetraacetaat
Formule: C₁₀H₁₂N₂Na₄O₈
Smeltpunt: > 300 °C
Dichtheid: 6,9 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150027
CAS nummer: 64-02-8
EINECS: 200-573-9
Merck Index: 12,03557
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3da8dc42f6b4faeb40c2c5e98f983e37d310fe21&vid=c925ac956246bbfd17c182e83b57c15a3b8edfa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 83 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR