Mierenzuur 100% ≥80%, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

20315.297EA 25.5 EUR
20315.297 20315.446 20315.366
Mierenzuur 100% ≥80%, TECHNICAL
Mierenzuur
Formule: HCO₂H
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003297
CAS nummer: 64-18-6
EINECS: 200-579-1
UN: 3412
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 79 - 81 %
n 20/D 1.369 - 1.372


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR