Lactic acid ≥90%, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

20366.293EA 101 EUR
20366.293 20366.464
Lactic acid ≥90%, TECHNICAL
Lactic acid
Formule: H₃CCH(OH)COOH
Kookpunt: 122 °C (20 hPa)
Smeltpunt: 53 °C
Dichtheid: 1,11…1.21 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004520
CAS nummer: 50-21-5
EINECS: 200-018-0

Order Now

Specification Test Results

Assay Min. 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR