Salpeterzuur 69%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Nitric acid solution

20425.242EA 49.8 EUR
20425.242 20425.297 20425.322 20425.420 20425.550
Salpeterzuur 69%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Salpeterzuur
Formule: HNO₃
Kookpunt: 120,5 °C (1013 hPa)
Smeltpunt: –42 °C
Dichtheid: 1,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 69.0 - 70.0 %
Appearance of solution Passes test Ph.Eur.
Identification (NO3) Passes test Ph.Eur.
Colouration Max. 10 APHA
Density (20/20) 1.384 - 1.416
Heavy metals (as Pb) Max. 0.2 ppm
Ignition residue (SO4) Max. 5 ppm
Silica Max. 0.2 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 1 ppm
Al (Aluminium) Max. 0.05 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.05 ppm
Mg (Magnesium) Max. 0.03 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.3 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sr (Strontium) Max. 0.01 ppm
Zn (Zinc) Max. 0.05 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR