Periodic acid ≥97%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: ortho-Periodic acid, Orthoperiodic acid

20592.124EA 39.2 EUR
20592.124 20592.181
Periodic acid ≥97%, GPR RECTAPUR®
Periodic acid
Formule: H₅IO₆
Smeltpunt: 122 °C
Dichtheid: 1,3875 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011344
CAS nummer: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1 (8),II
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 97 %
Ignition residue (SO4) Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR