Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

206231000 206235000 206230025
206231000.EA 51.6 EUR
206231000. 206235000. 206230025.
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder
Nickel aluminium alloy according to Raney

CAS No.: 12635-29-9

Formule: Ni:Al;₅₀:₅₀%
Smeltpunt: 1350 °C
Dichtheid: 3,46 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167392
CAS nummer: 12635-29-9
UN: 3208
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Grey to grey-black Powder
Aluminium (Al) 47.0 to 52.0 %
Nickel (Ni) 48.0 to 53.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR