Octylphosphonic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

20651.03 20651.06
20651.03EA 64.1 EUR
20651.03 20651.06
Octylphosphonic acid ≥98%
Octylphosphonic acid

Purity: 98% CAS No.: 4724-48-5 Molecular Formula: C8H19O3P Formula Weight: 194.21 Physical Form: Crystalline Melting Point: 100-102° Beilstein Reference: 1764242 MDL No.: MFCD00015841

Formule: C₈H₁₉O₃P
Smeltpunt: 100…102 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015841
CAS nummer: 4724-48-5
EINECS: 225-218-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR