Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.2 µm filter, packaged under nitrogen.

20837.320EA 48.8 EUR
20837.320 20837.360 20837.9010 20837.9030
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Methanol
Formule: CH₃OH
Kookpunt: 64,6 °C (1013 hPa)
Smeltpunt: –98 °C
Dichtheid: 0,7918 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
Bereik: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0003 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.05 %
Absorbance (220 nm) Max. 0.4
Absorbance (240 nm) Max. 0.15
Absorbance (260 nm) Max. 0.02
Transmittance (220 nm) Min. 45 %
Transmittance (240 nm) Min. 75 %
Transmittance (260 nm) Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR