Tetramethylammonium hydroxide 25% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

20932.AK 20932.K2
20932.AKEA 53.8 EUR
20932.AK 20932.K2
Tetramethylammonium hydroxide 25% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Tetramethylammonium hydroxide

Purity: 99.9999% CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Physical Form: Liquid Density: 1.016 Refractive Index: 1.3840 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Boiling Point: ca 102° Beilstein Reference: 3558708 Merck Reference: 14,9224 UN No.: 1835 MDL No.: MFCD00008280

Formule: C₄H₁₃NO
Kookpunt: ∼102 °C
Dichtheid: 1,016
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008280
CAS nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
Merck Index: 14,9224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR