Aluminium sulphate octadecahydrate, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trade termforalum, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

21068.290EA 0 EUR
21068.290
Aluminium sulphate octadecahydrate, TECHNICAL
Aluminium sulphate octadecahydrate
Formule: Al₂(SO₄)₃·18H₂O
Smeltpunt: 86 °C
Dichtheid: 1,69 g/cm³ (20 °C)
MDL Nummer: MFCD00149136
CAS nummer: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR