Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate

21094.233EA 27.7 EUR
21094.233 21094.290 21094.260
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
Ammonium iron(III) sulphate dodecahydrate
Formule: NH₄Fe(SO₄)₂·12H₂O
Kookpunt: 230 °C (1013 hPa)
Smeltpunt: 39…41 °C
Dichtheid: 1,71 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150004
CAS nummer: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insolubility in water Max. 50 ppm
Substances not precipitated by NH4OH Max. 0.1 %
Cl (Chloride) Max. 10 ppm
NO3 (Nitrate) Max. 100 ppm
Cu (Copper) Max. 20 ppm
Fe (II) (Iron) Max. 10 ppm
Mn (Manganese) Max. 50 ppm
Pb (Lead) Max. 20 ppm
Zn (Zinc) Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR