Ammoniak-oplossing 20%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ammoniumhydroxide, Waterig-ammoniakale oplossing

21180.242EA 25.2 EUR
21180.242 21180.297 21180.366 21180.446
Ammoniak-oplossing 20%, GPR RECTAPUR®
Ammoniak-oplossing
Formule: NH₃
Dichtheid: 0,92 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066650
CAS nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay 18 - 22 %
Density (20/4) 0.910 - 0.930
Evaporation residue Max. 0.02 %
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 20 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR