Ammoniak-oplossing 28%, AnalaR NORMAPUR® analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ammoniumhydroxide, Waterig-ammoniakale oplossing

21190.326EA 52 EUR
21190.326 21190.246 21190.292 21190.361
Ammoniak-oplossing 28%, AnalaR NORMAPUR® analytical reagent
Ammoniak-oplossing
Formule: NH₃
Kookpunt: 32…37,7 °C (1013 hPa)
Dichtheid: 0,898 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066650
CAS nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay 28.0 - 32.0 %
Colouration Max. 10 APHA
Density (20/4) 0.880 - 0.910
Evaporation residue Max. 10 ppm
Silica Max. 10 ppm
Substances reducing KMnO4 (as O) Max. 4 ppm
Cl (Chloride) Max. 0.5 ppm
CO3 (Carbonate) Max. 10 ppm
NO2 (Nitrite) Max. 2 ppm
PO4 (Phosphate) Max. 0.5 ppm
S (Sulphide) Max. 0.1 ppm
SO4 (Sulphate) Max. 2 ppm
Al (Aluminium) Max. 0.03 ppm
As (Arsenic) Max. 0.01 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Be (Beryllium) Max. 0.01 ppm
Ca (Calcium) Max. 0.3 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Hg (Mercury) Max. 0.01 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 2 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.05 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.02 ppm
Zr (Zirconium) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR