MPC series plastic couplings
Leverancier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MPC17004T03 MPC17004T39 MPC17004T39B25 MPC17006T03 MPC17006T03B25 MPC17006T39 MPC22004T03M MPC22004T03MB2 MPC22004T39M MPC22004T39MB2 MPC22006T03M MPC22006T03MB2 MPC22006T39M MPC22006T39MB2
212-0801EA 789 EUR
212-0801 212-0802 212-0803 212-0804 212-0805 212-0806 212-0807 212-0808 212-0809 212-0810 212-0811 212-0812 212-0813 212-0814
MPC series plastic couplings
Fittingen en connectoren
Couplings allow rapid disconnect and reconnect with one hand and without the need for tools.
Couplings are available in polysulphone (amber tint) or polycarbonate (purple tint) construction. Coupling inserts include seals constructed of clear silicone. All products meet USP Class VI criteria.
They feature an ergonomic thumb latch and an audible click when connected, assuring the user of a secure connection. Couplings can be rotated when connected, reducing the inconvenience of tube kinks. These items are sterilisable and reusable, yet economical enough to allow disposability.