NMR-monsterbuisjes, 5 mm, met nauwkeurigheidsglas, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
231700314 231700117 231700211
212-1682EA 94.6 EUR
212-1682 212-1680 SCOT231700211
NMR-monsterbuisjes, 5 mm, met nauwkeurigheidsglas, DURAN®
Buisjes NMR-buisjes
DURAN®, borosilicate glass 3.3, clear.
  • Storage and transport of measurement solutions
  • Designed for use in low to most mid field NMR spectrometers
  • Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution
DURAN® NMR tubes are available, according to requirement, in three accuracy classes. The correct tube can be selected depending on resonant frequency. These tubes are noteworthy for their close tolerances and accuracy, especially to their straightness, wall thickness and wall thicknessMore Product Information