Test tubes, with rim
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
88323 88324 88322 88317 88319 88325
KART88323EA 185 EUR
KART88323 KART88324 KART88322 KART88317 KART88319 212-6894
Test tubes, with rim
Buisjes Reageerbuisjes
PS or PP.
  • Disposable tubes with round or conical bottom
  • With rim
  • For laboratory use
  • Can be used in centrifuges
Certificaten: IVD certified.