Reageerbuizen zonder rand, dunwandig
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G031/01 G031/02 G031/03 G031/04 G031/05 G031/06 G031/08 G031/10 G031/09
212-7105EA 63.6 EUR
212-7105 212-7106 212-7107 212-7108 212-7032 212-7033 212-7034 212-0462 212-0565
Reageerbuizen zonder rand, dunwandig
Buisjes Reageerbuisjes
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certificaten: ISO 4142:2002