Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F199210001
212-7310EA 65 EUR
212-7310
Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Vijzels en stampers Stampers
Polypropylene tissue grinder efficiently homogenizes tissue and cells in microcentrifuge, centrifuge, and grinding tubes.
  • Polypropylene construction
  • Rounded end design
  • Tapered conical tip
The cone tip tapers from 8,2 mm (0,33") down to 2,9 mm (0,12") in a length of 19,8 mm (0,78"). It features a rounded end. Overall length is 21 cm (8¼"), with a 4,7 mm (³/₁₆") diameter shaft. Steam autoclavable at 121 °C (250 °F).
Order Now
Specificaties
Environmentally Preferable