Cluster tube racks, 1,2 ml, BioTube™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T101-1 T101-2 T101-3 T101-4 T101-6 T101-5
212-9556EA 196 EUR
212-9556 212-9557 212-9558 212-9559 SIMPT101-6 SIMPT101-5
Cluster tube racks, 1,2 ml, BioTube™
Buisjes Microbuisjes
These racks contain 96 removable PP tubes, either placed individually or in a 8×12 configuration.
  • Very good chemical resistance
  • Very good temperature resistance
  • Certified RNase, DNase, pyrogen- and DNA-free
  • Conform to SBS standards
  • Autoclavable
These racks are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA.
The BioTube™ system is designed in such a way that the 96-place rack, having a standard on-center spacing of tubes, also has aMore Product Information