Beakers, low form, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BPB0050P BPB0100P BPB0250P BPB1000P BPB0500P
213-0096EA 38 EUR
213-0096 213-0097 213-0098 213-0102 213-0100
Beakers, low form, Azlon®
Bekers
PP, highly translucent.
  • Tapered stacking style; space saving and easy to handle
  • Good chemical resistance
  • Autoclavable at +121 °C
Ideal for general laboratory use. Crisp, blue, easy to read indelible graduations. Precise non drip pouring spouts.
Certificaten: ISO 7056