Bekers, lage vorm, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BWN012 BWN018 BWN024 BWN030 BWN036
213-0138EA 33.7 EUR
213-0138 213-0139 213-0140 213-0141 213-0142
Bekers, lage vorm, Azlon®
Bekers
PTFE, opaque, rigid, non-porous.
  • Excellent chemical resistance - resists nearly all chemicals except fluorine and molten alkali metals
  • Can withstand 260 °C
  • Effective substitutes for platinum containers
Opgelet: It is NOT advisable to use PTFE beakers on a hotplate.