VWR®, Maatbekers met handvat, PP
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
213-0497EA 1.8 EUR
213-0497 213-0498 213-0499 213-0500 213-0501 213-0502 213-0503
VWR®, Maatbekers met handvat, PP
Bekers
Made of PP, translucent.
  • With printed graduations
  • With handle for better usage
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements