Ammonium sulphide 20% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Gevaar
21354.298EA 196 EUR
21354.298
Ammonium sulphide 20% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Ammonium sulphide
Formule: H₈N₂S
MW: 68.14 g/mol
Smeltpunt: -18
Dichtheid: 0.99
MDL Nummer: MFCD00010892
CAS nummer: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 (3, 6.1) II
Merck Index: 13,00560
Order Now
Specification Test Results
Assay (as (NH4)2S from sulphide) 17.0 - 23.0 %
Ignition residue (SO4) Max. 100 ppm
Cl (Chloride) Max. 0.02 %
CO3 (Carbonate) Max. 100 ppm