Holders for round bottom flasks, Flaskup™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F389512004 F389512006 F389512010 F389512012
214-1952EA 74.1 EUR
214-1952 214-1953 214-1954 214-1955
Holders for round bottom flasks, Flaskup™, SP Bel-Art
Steunen Kolfsteunen
Holders made from PP for round bottom flasks.
  • Inner wall has a slight taper to the open bottom
  • Allows variety of flask shapes and sizes to fit into one holder
  • Wide base keeps flasks stable
  • Side slots can be used as sight windows
  • Temperature resistant upto 121 °C
Bestelinformatie: Available as 3 per package, 8 packages per case and assortment packs.