Flasks, flat bottom, narrow or wide neck, with rim
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632417117100 632417117250 632417117500 632411117050 632417117954 632411117966 632411117958 632411103954 632411103050 632411103500 632411103940
214-3161EA 9.3 EUR
214-3161 214-3162 214-3163 KAVA632411117050 KAVA632417117954 KAVA632411117966 KAVA632411117958 KAVA632411103954 KAVA632411103050 KAVA632411103500 KAVA632411103940
Flasks, flat bottom, narrow or wide neck, with rim
Kolven Platbodemkolven
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.