Fosforpentoxide 98,0, AnalaR NORMAPUR® zA

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride

21411.296EA 284 EUR
21411.296 21411.230
Fosforpentoxide 98,0, AnalaR NORMAPUR® zA
Fosforpentoxide
Formule: P₂O₅
MW: 141.94 g/mol
Smeltpunt: 562
Dichtheid: 2.253 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011440
CAS nummer: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8 II
Merck Index: 13,07441

Order Now

Specification Test Results

Assay Min. 98.0 %
Insolubility in water Max. 50 ppm
Reducing substances (as P2O3) Max. 0.02 %
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 150 ppm
Cd (Cadmium) Max. 5 ppm
Cu (Copper) Max. 20 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 100 ppm
Mn (Manganese) Max. 5 ppm
Na (Sodium) Max. 0.02 %
Ni (Nickel) Max. 20 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR