Jars, wide mouth
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1973 1972 1974 1812 1933
215-0237EA 290 EUR
215-0237 215-0265 215-0278 SEMA1812 SEMA1933
Jars, wide mouth
Potjes Potjes voor algemeen gebruik
Jars, made of HDPE or PP, translucent. Resistant to shape, solid design, optionally with close lid.
  • Good chemical resistance
  • Good temperature resistance
  • Lightweight
  • Lids optionally available
Very cost-effective, also suitable as a disposable cup or packaging.