Decanteerflessen met zijtubus
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1531/08D 1531/10D 1531/12D
215-0349EA 527 EUR
215-0349 BIBB1531/10D 215-0350
Decanteerflessen met zijtubus
Flessen Doseerflessen
PYREX®, borosilicate glass, without stoppers.
  • Excellent chemical resistance
  • Very good temperature resistance
Clear with white labelling area and unground neck to accept rubber stoppers. Glass tubular side arm for attachment of 12 mm bore flexible tubing.