Laboratoriumflessen, smalle hals, met schroefdraad, amberkleurig glas, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
218061409 218061709 218062405 218063607 218064406 218065402 218066304 218067309 218068605 218068802 218069104 218062902 218065102 218066904 218061458 218061758 218062454 218062951 218063656 218064455 218065151 218065451 218066353 218066953 218067358 218068654 218060807 218060856
215-1572EA 22.1 EUR
215-1572 215-1573 215-1574 215-1575 215-1576 215-1577 215-1578 215-1579 215-1580 215-1581 215-1582 215-2434 215-2435 215-2436 215-4487 215-3793 215-3794 215-3796 215-3797 215-3798 215-3804 215-3805 215-3799 215-3806 215-3807 215-3808 215-3296 215-3215
Laboratoriumflessen, smalle hals, met schroefdraad, amberkleurig glas, DURAN®
Flessen Glazen flessen
DURAN®, borosilicaatglas 3.3, bruin, zonder of met gietring en schroefdop.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Purely external application of the colour ensures unchanged DURAN® properties on the interior
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
  • 10 ml bottles do not conform with ISO 4796-1 standard; specially shaped glass edge for improved pouring out (so that an additional
More Product Information