Flessen, middelgrote hals, HDPE, met schroefdop, serie 307
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
307-770066 307-770067 307-770070 307-770073
215-2331EA 21.1 EUR
215-2331 215-2332 215-2333 215-2334
Flessen, middelgrote hals, HDPE, met schroefdop, serie 307
Flessen Kunststof flessen
HDPE, rond, natuurlijke kleur, met sluiting, met schaalverdeling.
  • Embossed contents scale
  • Easy to fill
  • Easy to clean
  • Leakproof
  • Multi-purpose bottles for laboratories and industry
  • The neck opening means that the bottles are easy to fill and clean
  • Halal certified
Polypropylene screw closure.