Flessen, brede hals, vierkant zonder schroefdop
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
310-71138 310-71140 310-71143 310-71144 310-73094 310-73095 310-73096 310-73097 310-73624 310-76398 310-79206 310-79207 310-79209 310-79210 310-79211 310-79212 310-79213 310-79214 310-84193
215-3375EA 2.5 EUR
215-3375 215-2361 215-2364 215-2365 215-3604 215-3603 215-3380 215-2362 215-2363 215-3601 215-3362 215-3363 215-3364 215-2371 215-3365 215-2372 215-2373 215-2374 215-2370
Flessen, brede hals, vierkant zonder schroefdop
Flessen Kunststof flessen
HDPE, transparant, of HDPE, wit, zonder dop.
  • Square cross-section
  • Recessed labelling field
  • Three different types of closures available
  • Leakproof
  • Suitable for food
  • Wide neck opening for easy filling and cleaning
  • Halal certified
Ideal for liquids, pastes, granulates, and powders. With UN approval for solids (except nominal capacities 50 ml and 350 ml). The approvals are only valid with the closures offered here.