Flessen, smalle hals, vierkant, HDPE, met schroefdraad
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
310-71149 310-82553 310-83970 310-83971 310-83972 310-83973 310-83991 310-83992 310-83994 310-84000
215-1405EA 8.1 EUR
215-1405 215-3689 215-2366 215-2367 215-2368 215-2369 215-3354 215-3355 215-3353 215-3352
Flessen, smalle hals, vierkant, HDPE, met schroefdraad
Flessen Kunststof flessen
HDPE. Square. Natural or white colour.
  • Square cross-section
  • Recessed labelling field
  • Leakproof
  • Bottles are suitable for food
  • Ideal for liquids
  • Halal certified
Five different types of closures available.