Flessen, smalle hals, vierkant, "Clear Grip"
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
310-86456 310-86457 310-86458 310-86459
215-2879EA 599 EUR
215-2879 215-2880 215-2881 215-2882
Flessen, smalle hals, vierkant, "Clear Grip"
Flessen Kunststof flessen
PP, zeer transparant,
  • Low weight
  • Stiffness
  • Impact resistance
  • Easily recyclable
  • Leakproof
  • Suitable for food
  • Halal certified
Good transparency and glaze (ideal for product presentations). Outstanding chemical resistance and thermal stability, balanced stiffness, and impact resistance.